missing translation for 'onlineSavingsMsg'
Learn More

n-Octyltrichlorosilane, 97%

Product Code. 11480188
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480188 500 g 500g
11460188 25 g 25g
11470188 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480188 Supplier Thermo Scientific Alfa Aesar Supplier No. B23753.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5283-66-9 | C8H17Cl3Si | 247.659 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5283-66-9
Molecular Formula C8H17Cl3Si
Molecular Weight (g/mol) 247.659
MDL Number MFCD00000488
InChI Key RCHUVCPBWWSUMC-UHFFFAOYSA-N
Synonym trichloro octyl silane, n-octyltrichlorosilane, octyltrichlorosilane, silane, trichlorooctyl, trichloro-n-octylsilane, unii-jv18i1wcu8, jv18i1wcu8, silane, octyltrichloro, octyltrichlorosilane un1801 corrosive
PubChem CID 21354
IUPAC Name trichloro(octyl)silane
SMILES CCCCCCCC[Si](Cl)(Cl)Cl

Specifications

Boiling Point 225°C to 227°C
Quantity 500 g
UN Number UN1801
Beilstein 1744230
Flash Point 96°C (204°F)
Formula Weight 247.67
Percent Purity 97%
Odor Irritating
Refractive Index 1.447
Sensitivity Moisture sensitive
Density 1.073
Chemical Name or Material n-Octyltrichlorosilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.