missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Octylbicycloheptenedicarboximide, TRC
Specifications
Specifications
| Chemical Name or Material | MGK 264 |
| CAS | 113-48-4 |
| Molecular Formula | C17 H25 N O2 |
| Quantity | 250 mg |
| Synonym | 4,7-Methano-1H-isoindole-1,3(2H)-dione','2-(2-ethylhexyl)-3a,4,7,7a-tetrahydro-, 5-Norbornene-2,3-dicarboximide','N-(2-ethylhexyl)- (6CI,7CI,8CI), 2-(2-Ethylhexyl)-3a,4,7,7a-tetrahydro-4,7-methano-1H-isoindole-1,3(2H)-dione, Carboximide, Dicarboximide, ENT 8184, MGK 264, N-(2-Ethylhexyl)-5-norbornene-2,3-dicarboximide, N-(2-Ethylhexyl)bicyclo-(2,2,1)-5-heptene-2,3-dicarboximide, NSC 36678, NSC 406879, Octacide 264, Octylbicycloheptenedicarboximide, Synepirin 222, Synergist 264, Van Dyk 264, Zengxiaoan |
| InChI Formula | InChI=1S/C17H25NO2/c1-3-5-6-11(4-2)10-18-16(19)14-12-7-8-13(9-12)15(14)17(18)20/h7-8,11-15H,3-6,9-10H2,1-2H3/t11?,12-,13+,14?,15? |
| SMILES | CCCCC(CC)CN1C(=O)C2[C@@H]3C[C@@H](C=C3)C2C1=O |
| IUPAC Name | N-(2-Ethylhexyl)-5-norbornen-2,3-dicarboximide |
| Molecular Weight (g/mol) | 275.39 |
| Formula Weight | 275.1885 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?