missing translation for 'onlineSavingsMsg'
Learn More

n-Octylbenzene, 99%

Product Code. 11410103
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410103 25 g 25g
11420103 100 g 100g
11430103 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410103 Supplier Thermo Scientific Alfa Aesar Supplier No. A12138.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2189-60-8 | C14H22 | 190.33 g/mol

n-octylbenzene is used as a solvent to study the self-assembly of derivatives of tetrathiafulvalene (TTF) on graphite and their visualization by scanning tunnelling microscopy (STM). It is also involved in the study the effects and substitution of these compounds. It is also used to prepare charge-transfer complexes with fluoranil and 1,3,5-trinitrobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-octylbenzene is used as a solvent to study the self-assembly of derivatives of tetrathiafulvalene (TTF) on graphite and their visualization by scanning tunnelling microscopy (STM). It is also involved in the study the effects and substitution of these compounds. It is also used to prepare charge-transfer complexes with fluoranil and 1,3,5-trinitrobenzene.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2189-60-8
Molecular Formula C14H22
Molecular Weight (g/mol) 190.33
MDL Number MFCD00009564
InChI Key CDKDZKXSXLNROY-UHFFFAOYSA-N
Synonym n-octylbenzene, 1-phenyloctane, benzene, octyl, phenyloctane, octane, 1-phenyl, octyl-benzene, 1-octylbenzene, octyl benzene, 4-octylbenzene, acmc-1cplg
PubChem CID 16607
IUPAC Name octylbenzene
SMILES CCCCCCCCC1=CC=CC=C1

Specifications

Melting Point -36°C
Density 0.854
Boiling Point 264°C to 265°C
Flash Point 107°C (224°F)
Odor Mild
Refractive Index 1.484
Quantity 25 g
Beilstein 1906253
Solubility Information Immiscible with water.
Formula Weight 190.33
Percent Purity 99%
Chemical Name or Material n-Octylbenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.