missing translation for 'onlineSavingsMsg'
Learn More

N-Octyl Nortadalafil, TRC

Product Code. 30137510
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137510 10 mg 10mg
30135345 25 mg 25mg
30011200 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137510 Supplier Toronto Research Chemicals Supplier No. TRCO24135010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1173706-35-8
Molecular Formula C29 H33 N3 O4
Molecular Weight (g/mol) 487.59
Synonym Pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, 6-(1,3-benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-2-octyl-, (6R,12aR)-, N-Octylnortadalafil
IUPAC Name (2R,8R)-2-(1,3-benzodioxol-5-yl)-6-octyl-3,6,17-triazatetracyclo[8.7.0.0^{3,8}.0^{11,16}]heptadeca-1(10),11,13,15-tetraene-4,7-dione
SMILES CCCCCCCCN1CC(=O)N2[C@H](Cc3c([nH]c4ccccc34)[C@H]2c5ccc6OCOc6c5)C1=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C29H33N3O4/c1-2-3-4-5-6-9-14-31-17-26(33)32-23(29(31)34)16-21-20-10-7-8-11-22(20)30-27(21)28(32)19-12-13-24-25(15-19)36-18-35-24/h7-8,10-13,15,23,28,30H,2-6,9,14,16-18H2,1H3/t23-,28-/m1/s1
Formula Weight 487.2471
Percent Purity >95%
Chemical Name or Material N-Octyl Nortadalafil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.