missing translation for 'onlineSavingsMsg'
Learn More

N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, 98%

Product Code. 15507049 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15507049 25 g 25g
15597039 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15507049 Supplier Thermo Scientific Acros Supplier No. 459700250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15178-76-4 | C13H29NO3S | 279.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15178-76-4
Molecular Formula C13H29NO3S
Molecular Weight (g/mol) 279.44
InChI Key QZRAABPTWGFNIU-UHFFFAOYSA-N
Synonym octyl sulfobetaine, 3-dimethyl-octylazaniumyl propane-1-sulfonate, n-octyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-n,n-dimethyloctylammonio propanesulfonate inner salt, 3-dimethyl octyl ammonio propane-1-sulfonate, 3-n,n-dimethyloctylammonio propanesulfonate solution, n-octyl-n,n-dimethyl-3-ammonio-1-propanesulfonate sb-8, n-octylsulfobetaine, zwittergent 3-08
PubChem CID 3084219
IUPAC Name 3-[dimethyl(octyl)azaniumyl]propane-1-sulfonate
SMILES CCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]

Specifications

Melting Point 217.0°C
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Solubility Information (10% in water) Clear solution
Formula Weight 279.44
Percent Purity 98%
Chemical Name or Material N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
compliance-icons
Product Identifier
  • N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.