missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl-β-D-glucopyranoside (White Crystalline Powder), Fisher BioReagents

Product Code. p-27452
Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
Packaging:
Amber Poly Bottle
Poly Micro Tube
Unit Size:
1g
25g
500mg
5g
This item is not returnable. View return policy

Product Code. 10214873

Brand: Fisher BioReagents BP585500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
n-Octyl-β-D-Glucopyranoside is a highly purified, nonionic detergent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29836-26-8
Molecular Formula C14H28O6
Molecular Weight (g/mol) 292.37
MDL Number MFCD00063288
InChI Key HEGSGKPQLMEBJL-RKQHYHRCSA-N
PubChem CID 62852
SMILES CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Absorbance 0.3 max. at 260nm
Color White
Physical Form Powder/Solid
Melting Point 105°C
Quantity 500 mg
Packaging Poly Micro Tube
Assay Percent Range ≥98 %
Specific Rotation -28.0 to -32.0° (+ or -)
Merck Index 15, 6857
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol
Formula Weight 292.4
Chemical Name or Material n-Octyl-β-D-Glucopyranoside
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.