missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl-β-D-glucopyranoside (White Crystalline Powder), Fisher BioReagents

Product Code. 10214873
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
Packaging:
Amber Poly Bottle
Poly Micro Tube
Unit Size:
1g
25g
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10214873 500 mg Poly Micro Tube 500mg
10071693 1 g Poly Micro Tube 1g
10326843 5 g Amber Poly Bottle 5g
10071073 25 g Amber Poly Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10214873 Supplier Fisher BioReagents Supplier No. BP585500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
n-Octyl-β-D-Glucopyranoside is a highly purified, nonionic detergent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29836-26-8
Molecular Formula C14H28O6
Molecular Weight (g/mol) 292.37
MDL Number MFCD00063288
InChI Key HEGSGKPQLMEBJL-RKQHYHRCSA-N
PubChem CID 62852
SMILES CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Absorbance 0.3 max. at 260nm
Color White
Physical Form Powder/Solid
Melting Point 105°C
Quantity 500 mg
Packaging Poly Micro Tube
Assay Percent Range ≥98 %
Specific Rotation -28.0 to -32.0° (+ or -)
Merck Index 15, 6857
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol
Formula Weight 292.4
Chemical Name or Material n-Octyl-β-D-Glucopyranoside
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.