missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl-beta-D-glucopyranoside, 98+%

Product Code. 15488719
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 15488719

Brand: Thermo Scientific Alfa Aesar J67390.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29836-26-8 | C14H28O6 | 292.37 g/mol

It is used for the solubilization and reconstitution of membrane-bound proteins. The high critical micelle concentration (0.7%) of n-octylglucoside facilitates ready removal from final protein extracts by dialysis or gel filtration. n-Octylglucoside can be used in 2D electrophoresis and to improve selectivity of immunoprecipitation of phosphotyrosine modified proteins. Also useful in 2D electrophoresis and in an immunoprecipitation buffer for the immunopurification of phospho-tyrosine peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for the solubilization and reconstitution of membrane-bound proteins. The high critical micelle concentration (0.7%) of n-octylglucoside facilitates ready removal from final protein extracts by dialysis or gel filtration. n-Octylglucoside can be used in 2D electrophoresis and to improve selectivity of immunoprecipitation of phosphotyrosine modified proteins. Also useful in 2D electrophoresis and in an immunoprecipitation buffer for the immunopurification of phospho-tyrosine peptides.

Solubility
Soluble in DMSO, methanol and water.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Store under inert gas. Store in refrigerator at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29836-26-8
Molecular Formula C14H28O6
Molecular Weight (g/mol) 292.37
MDL Number MFCD00063288
InChI Key HEGSGKPQLMEBJL-RKQHYHRCSA-N
Synonym octyl-beta-d-glucopyranoside, b-octylglucoside, octyl beta-d-glucopyranoside, octyl beta-d-glucoside, octyl glucoside, n-octyl-beta-d-glucoside, octyl b-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol, unii-v109wut6rl, n-octyl glucoside
PubChem CID 62852
SMILES CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 103°C to 104°C
Color White
Quantity 5 g
Beilstein 84118
Sensitivity Very Hygroscopic
Solubility Information Soluble in DMSO,methanol and water.
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol
Formula Weight 292.37
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material n-Octyl-beta-D-glucopyranoside
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.