missing translation for 'onlineSavingsMsg'
Learn More

N-Octyl 4-Hydroxybenzoate, TRC

Product Code. 30425596
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
500mg
Product Code. Quantity unitSize
30425596 1 g 1g
30410457 100 mg -
30425368 500 mg 500mg
3 options
This item is not returnable. View return policy

Product Code. 30425596

Brand: Toronto Research Chemicals TRCO1484731G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1219-38-1
Formula Weight 250.1569
InChI Formula InChI=1S/C15H22O3/c1-2-3-4-5-6-7-12-18-15(17)13-8-10-14(16)11-9-13/h8-11,16H,2-7,12H2,1H3
IUPAC Name octyl 4-hydroxybenzoate
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.33
SMILES CCCCCCCCOC(=O)c1ccc(O)cc1
Synonym Benzoic acid, 4-hydroxy-, octyl ester (9CI, ACI), Benzoic acid, p-hydroxy-, octyl ester (6CI, 7CI, 8CI), n-Octyl 4-hydroxybenzoate, n-Octyl p-hydroxybenzoate, NSC 309820, Octyl 4-hydroxybenzoate, Octyl p-hydroxybenzoate, Octyl paraben, p-Hydroxybenzoic acid octyl ester
Chemical Name or Material N-Octyl 4-Hydroxybenzoate

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.