missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl 4-hydroxybenzoate, 98%

Product Code. 11498526
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498526 25 g 25g
11478720 5 g 5g
11408536 100 g 100g
11418536 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11498526 Supplier Thermo Scientific Alfa Aesar Supplier No. A15964.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1219-38-1 | C15H22O3 | 250.34 g/mol

n-Octyl 4-hydroxybenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Octyl 4-hydroxybenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in alcohol, phenoxy ethanol and propylene glycol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1219-38-1
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.34
MDL Number MFCD00016482
InChI Key RIKCMEDSBFQFAL-UHFFFAOYSA-N
Synonym octylparaben, n-octyl 4-hydroxybenzoate, octyl paraben, octyl p-hydroxybenzoate, p-hydroxybenzoic acid octyl ester, benzoic acid, 4-hydroxy-, octyl ester, p-oxybenzoesaureoctylester, 4-hydroxybenzoic acid n-octyl ester, unii-4uvk431k6b, p-oxybenzoesaureoctylester german
PubChem CID 14642
IUPAC Name octyl 4-hydroxybenzoate
SMILES CCCCCCCCOC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 50°C to 53°C
Quantity 25 g
Solubility Information Soluble in alcohol,phenoxy ethanol and propylene glycol. Insoluble in water.
Formula Weight 250.34
Percent Purity 98%
Chemical Name or Material n-Octyl 4-hydroxybenzoate
compliance-icons
Product Identifier
  • n-Octyl 4-hydroxybenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Octyl 4-hydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.