missing translation for 'onlineSavingsMsg'
Learn More

N-Octanoic Acid, Extra Pure, SLR, Fisher Chemical™

Product Code. 10617612 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
Plastic container
Unit Size:
2.5 L
25 L
500mL
This item is not returnable. View return policy

Product Code. 10617612

Brand: Fisher Chemical O/0030/08

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 124-07-2
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number 4429
InChI Key WWZKQHOCKIZLMA-UHFFFAOYSA-N
Synonym caprylic acid, n-octanoic acid, octylic acid, n-caprylic acid, octoic acid, n-octylic acid, n-octoic acid, neo-fat 8, 1-heptanecarboxylic acid, enantic acid
PubChem CID 379
ChEBI CHEBI:28837
IUPAC Name octanoic acid
SMILES CCCCCCCC(=O)O

Specifications

Melting Point 16°C
Color Yellow
Boiling Point 237°C
Packaging Amber glass bottle
Quantity 500 mL
Vapor Density 5
Formula Weight 144.21g/mol
Physical Form Liquid
compliance-icons
Product Identifier
  • Octanoic acid
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.