missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Octane-d18, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | n-Octane (D18) |
| CAS | 17252-77-6 |
| Molecular Formula | C8 D18 |
| Quantity | 5 mg |
| Synonym | Octane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-d18 (ACI), Octane-d18 (6CI','7CI','8CI','9CI), 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Octadecadeuteriooctane, n-Octane-2H18, n-Octane-d18, Perdeuterooctane, Octane-D18 |
| InChI Formula | InChI=1S/C8H18/c1-3-5-7-8-6-4-2/h3-8H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H] |
| IUPAC Name | 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-octadecadeuteriooctane |
| Molecular Weight (g/mol) | 132.34 |
| Formula Weight | 132.2538 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction