missing translation for 'onlineSavingsMsg'
Learn More

n-Octane-d{18}, 99% (Isotopic)

Product Code. 11374428
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11374428

Brand: Thermo Scientific Alfa Aesar 042339.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 17252-77-6 | C8H18 | 132.342 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17252-77-6
Molecular Formula C8H18
Molecular Weight (g/mol) 132.342
MDL Number MFCD00037626
InChI Key TVMXDCGIABBOFY-VAZJTQEUSA-N
Synonym octane-d18, 2h18 octane, n-octane-d18, 2 h?? octane, octane-d18, 98 atom % d, n-octane-d isotopic 1g
PubChem CID 519375
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-octadecadeuteriooctane
SMILES CCCCCCCC

Specifications

Physical Form Liquid
Density 0.815 g/mL
Flash Point 15.6°C (60°F)
Quantity 1 g
Melting Point -57°C
Boiling Point 126°C
UN Number UN1262
Formula Weight 132.34
Percent Purity 99%
Chemical Name or Material n-Octane-d{18}, Isotopic

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.