Learn More
n-Octadecylphosphonic acid, 97%
Description
n-Octadecylphosphonic acid is used in thermal paper for receipts, adding machines and tickets. It acts as surfactant, emulsifier, dispersants and chelating agents. Further, it is a long chain phosphonic acid compound utilized for proteomics research. In addition to this, it is used to prepare modified mesoporous magnetic nano particles through Lewis acid/base interaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 92°C to 98°C |
| Color | White |
| Linear Formula | CH3(CH2)17P(O)(OH)2 |
| Quantity | 5 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 334.48 |
| Percent Purity | 97% |
| Physical Form | Crystalline |
| Chemical Name or Material | n-Octadecylphosphonic acid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.