missing translation for 'onlineSavingsMsg'
Learn More

n-Octadecyl mercaptan, 96%

Product Code. 10173541 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10173541 25 g Glass bottle 25g
10317241 100 g Glass bottle 100g
10544314 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10173541 Supplier Thermo Scientific Acros Supplier No. 354980250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2885-00-9 | C18H38S | 286.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2885-00-9
Molecular Formula C18H38S
Molecular Weight (g/mol) 286.56
MDL Number MFCD00004886
InChI Key QJAOYSPHSNGHNC-UHFFFAOYSA-N
Synonym octadecanethiol, 1-octadecanethiol, stearyl mercaptan, n-octadecyl mercaptan, octadecyl mercaptan, 1-mercaptooctadecane, octadecanthiol, n-octadecanethiol, octadecanethol, stearylmercaptan
PubChem CID 17905
IUPAC Name octadecane-1-thiol
SMILES CCCCCCCCCCCCCCCCCCS

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 25.0°C to 35.0°C
Color White
Boiling Point 366.0°C
Flash Point 185°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)16CH2SH
Refractive Index 1.4645
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents
Formula Weight 286.56
Percent Purity 96%
Physical Form Crystalline Solid
Chemical Name or Material n-Octadecyl mercaptan
Show More Show Less
compliance-icons
Product Identifier
  • n-Octadecyl mercaptan
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.