missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ n-Octadecane, 99.5+%

Product Code. 11475240
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475240 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11475240 Supplier Alfa Aesar™ Supplier No. 044512.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Specifications

CAS 593-45-3
Melting Point 28°C
Color White
Density 0.782g/mL
Boiling Point 317°C
Flash Point 166°C (330°F)
Odor Odorless
Molecular Formula C18H38
Refractive Index 1.4390
MDL Number MFCD00009007
Quantity 5 g
Synonym n-octadecane, octadecan, unii-n102p6haiu, n102p6haiu, octadecane, octadecane, analytical standard, octadecane, n, oktadekan, ccris 681, stearyl group
InChI Key RZJRJXONCZWCBN-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCCC
IUPAC Name octadecane
Molecular Weight (g/mol) 254.502
PubChem CID 11635
ChEBI CHEBI:32926
Formula Weight 254.5
Percent Purity ≥99.5%
Physical Form Crystalline Mass/Melt
Chemical Name or Material n-Octadecane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.