missing translation for 'onlineSavingsMsg'
Learn More

n-Octacosane, 99%, Thermo Scientific Chemicals

Product Code. 11368968
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-02-4
Molecular Formula C28H58
Molecular Weight (g/mol) 394.77
MDL Number MFCD00009355
InChI Key ZYURHZPYMFLWSH-UHFFFAOYSA-N
Synonym n-octacosane, n-octcosane, unii-vff49836p8, ch3-ch2 26-ch3, octacosane, analytical standard, ccris 680, octacosane, n, octacosane, heptacosane, methyl, n-octacosane 10g
PubChem CID 12408
ChEBI CHEBI:32943
IUPAC Name octacosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 61°C to 63°C
Boiling Point 440°C
Flash Point 227°C (441°F)
Quantity 10 g
Solubility Information Soluble in chloroform,acetone,benzene,and toluene. Insoluble in water.
Formula Weight 394.77
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material n-Octacosane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'