missing translation for 'onlineSavingsMsg'
Learn More

N,O-Dimethylhydroxylamine Hydrochloride, TRC

Product Code. 30387106
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30387106 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387106 Supplier Toronto Research Chemicals Supplier No. TRCD473400100G
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material N,O-Dimethylhydroxylamine Hydrochloride
CAS 6638-79-5
Molecular Formula C2 H7 N O . H Cl
Quantity 100 g
Synonym N-Methoxymethylamine Hydrochloride, N-Methyl-N-methoxyamine Hydrochloride, O-Methyl Hydroxy(methyl)amine Hydrochloride
InChI Formula InChI=1S/C2H7NO.ClH/c1-3-4-2;/h3H,1-2H3;1H
SMILES Cl.O(NC)C
IUPAC Name N-methoxymethanamine;hydrochloride
Molecular Weight (g/mol) 97.54
Formula Weight 97.0294
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.