missing translation for 'onlineSavingsMsg'
Learn More

N,O-Dimethylhydroxylamine hydrochloride, 98%

Product Code. 10316420 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10316420 100 g Glass bottle 100g
10570741 5 g Glass bottle 5g
10030790 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10316420 Supplier Thermo Scientific Acros Supplier No. 116341000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6638-79-5 | C2H8ClNO | 97.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6638-79-5
Molecular Formula C2H8ClNO
Molecular Weight (g/mol) 97.54
MDL Number MFCD00012485
InChI Key USZLCYNVCCDPLQ-UHFFFAOYSA-N
Synonym n,o-dimethylhydroxylamine hydrochloride, n,o-dimethylhydroxylamine hcl, n-methoxymethylamine hydrochloride, methanamine, n-methoxy-, hydrochloride, o,n-dimethylhydroxylamine hydrochloride, n-methoxymethanamine hydrochloride, o,n-dimethyl-hydroxylamine hydrochloride, methoxy methyl amine hydrochloride, n,o-dimethylhydroxyamine hydrochloride, n,o-dimethylhydroxylaminehydrochloride
PubChem CID 81138
SMILES [H+].[Cl-].CNOC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 112.0°C to 116.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3ONHCH3·HCl
Quantity 100 g
Beilstein 04, 534
Fieser 11,201; 12,205; 15,144
Solubility Information (10% in water) Clear
IUPAC Name hydrogen methoxy(methyl)amine chloride
Formula Weight 97.55
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material N, O-Dimethylhydroxylamine hydrochloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • N,O-Dimethylhydroxylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.