missing translation for 'onlineSavingsMsg'
Learn More

N,O-Didesmethyltramadol Hydrochloride, Mikromol™

Product Code. 30050947
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
30050947 100 mg 100mg
30050225 25 mg 25mg
2 options
This item is not returnable. View return policy

Product Code. 30050947

Brand: Mikromol MM0007.08

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N,O-Didesmethyltramadol Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,O-Didesmethyltramadol Hydrochloride
CAS 333338-16-2
Analyte or Component Names N,O-Didesmethyltramadol Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C14 H21 N O2 . Cl H
Certifications/Compliance ISO 17025
Synonym N,O-Didesmethyltramadol Hydrochloride, Guaiacol Imp. H (EP)
InChI Formula InChI=1S/C14H21NO2.ClH/c1-15-10-12-5-2-3-8-14(12,17)11-6-4-7-13(16)9-11;/h4,6-7,9,12,15-17H,2-3,5,8,10H2,1H3;1H/t12-,14+;/m1./s1
SMILES Cl.CNC[C@H]1CCCC[C@]1(O)c2cccc(O)c2
IUPAC Name 3-[(1R,2R)-1-hydroxy-2-(methylaminomethyl)cyclohexyl]phenol;hydrochloride
Molecular Weight (g/mol) 271.78
Quantity 100 mg
Formula Weight 271.1339 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.