missing translation for 'onlineSavingsMsg'
Learn More

N,O-Dibenzoyl-L-tyrosine (>90%), TRC

Product Code. 30377892
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377892 10 g 10g
30371143 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30377892 Supplier Toronto Research Chemicals Supplier No. TRCD42267010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14325-35-0
Molecular Formula C23H19NO5
Molecular Weight (g/mol) 389.4
Synonym N-Benzoyl-L-Tyrosine Benzoate (Ester), L-N-Benzoyltyrosine Benzoate (Ester), Tiropramide Impurity A
SMILES CC(=O)O[C@@H]1C(OC(C)=O)C(OC(C)=O)[C@@H]2OC1CO2

Specifications

InChI Formula InChI=1S/C23H19NO5/c25-21(17-7-3-1-4-8-17)24-20(22(26)27)15-16-11-13-19(14-12-16)29-23(28)18-9-5-2-6-10-18/h1-14,20H,15H2,(H,24,25)(H,26,27)
Quantity 10 g
Shipping Condition Room Temperature
Chemical Name or Material N,O-Dibenzoyl-L-tyrosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.