missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)hydroxylamine, 97%

Product Code. 10444653 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10444653 1 g Glass bottle 1g
10451831 5 g Glass bottle 5g
10681555 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10444653 Supplier Thermo Scientific Acros Supplier No. 215980010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22737-37-7 | C6H19NOSi2 | 177.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22737-37-7
Molecular Formula C6H19NOSi2
Molecular Weight (g/mol) 177.39
MDL Number MFCD00008266
InChI Key ZAEUMMRLGAMWKE-UHFFFAOYSA-N
Synonym n,o-bis trimethylsilyl hydroxylamine, 1,1,1-trimethyl-n-trimethylsilyl oxy silylamine, silanamine, 1,1,1-trimethyl-n-trimethylsilyl oxy, 2,2,5,5-tetramethyl-3-oxa-4-aza-2,5-disilahexane, acmc-209fyl, hydroxylamine, 2tms derivative, n,o-bis-trimethylsilylhydroxylamine, n,o-bis trimethylsilyl hydroxylamin, n,o-bis-trimethylsilyl hydroxylamine
PubChem CID 89815
SMILES C[Si](C)(C)NO[Si](C)(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.8300g/mL
Boiling Point 78.0°C to 80.0°C (100.0 mmHg)
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SiONHSi(CH3)3
Refractive Index 1.4112
Quantity 1 g
Fieser 06,58; 08,523; 09,512; 06,183; 10,142; 12,183; 13,110; 16,128
Specific Gravity 0.83
IUPAC Name 2,2,5,5-tetramethyl-3-oxa-4-aza-2,5-disilahexane
Formula Weight 177.4
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N, O-Bis(trimethylsilyl)hydroxylamine, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,O-Bis(trimethylsilyl)hydroxylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.