missing translation for 'onlineSavingsMsg'
Learn More

N-Nornuciferine, MedChemExpress

Product Code. 16489258
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489258 10 mg 10mg
16461678 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16489258 Supplier Medchem Express Supplier No. HYN2129
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress N-Nornuciferine is an aporphine alkaloid in lotus leaf that significantly inhibits CYP2D6 with IC50 and Ki of 3.76 and 2.34 μM, respectively.

N-Nornuciferine is an aporphine alkaloid in lotus leaf that significantly inhibits CYP2D6 with IC50 and Ki of 3.76 and 2.34 μM, respectively.

Specifications

Chemical Name or Material N-Nornuciferine
CAS 4846-19-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H19NO2
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (355.43 mM; Need ultrasonic)
SMILES COC1=C(OC)C2=C3C(CCN[C@]3([H])CC4=CC=CC=C24)=C1
Molecular Weight (g/mol) 281.35
Formula Weight 281.35
Percent Purity 99.18%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.