missing translation for 'onlineSavingsMsg'
Learn More

n-Nonylbenzene, 97%

Product Code. 11439030
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. Quantity unitSize
11439030 50 g 50g
11429030 10 g 10g
11449030 250 g 250g
3 options
This item is not returnable. View return policy

Product Code. 11439030

Brand: Thermo Scientific Alfa Aesar A12957.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1081-77-2 | C15H24 | 204.357 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1081-77-2
Molecular Formula C15H24
Molecular Weight (g/mol) 204.357
MDL Number MFCD00009575
InChI Key LIXVMPBOGDCSRM-UHFFFAOYSA-N
Synonym 1-phenylnonane, benzene, nonyl, n-nonylbenzene, nonane, 1-phenyl, 1-nonylbenzene, unii-58ak68ov26, benzene, c6-12-alkyl derivs., n-nonyl benzene, phenyl-nonane, c6-c12 alkylbenzene
PubChem CID 14126
IUPAC Name nonylbenzene
SMILES CCCCCCCCCC1=CC=CC=C1

Specifications

Density 0.858
Boiling Point 281°C to 283°C
Flash Point >110°C (230°F)
Refractive Index 1.483
Quantity 50 g
Beilstein 1907173
Formula Weight 204.36
Percent Purity 97%
Chemical Name or Material n-Nonylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.