missing translation for 'onlineSavingsMsg'
Learn More

n-Nonanoic Acid, TRC

Product Code. 30471223
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30471223 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30471223

Brand: Toronto Research Chemicals undefined

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nonanoic acid (Pelargonic acid)
CAS 112-05-0
Purity Grade Notes GC
Molecular Formula C9 H18 O2
Quantity 5 g
Synonym Nonanoic acid (7CI','8CI','9CI','ACI), 1-Octanecarboxylic acid, Emerion W 90PA, Emery 1202, Emery 1203, Grantrico, n-Nonanoic acid, n-Nonoic acid, n-Nonylic acid, n-Pelargonic acid, Nonoic acid, Nonylic acid, NSC 62787, NSC 65450, NSC 65455, Pelargonic acid, Prifrac 2914, Thinex
InChI Formula InChI=1S/C9H18O2/c1-2-3-4-5-6-7-8-9(10)11/h2-8H2,1H3,(H,10,11)
SMILES CCCCCCCCC(=O)O
IUPAC Name nonanoic acid
Molecular Weight (g/mol) 158.24
Formula Weight 158.1307
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.