missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Nonanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Nonanoic acid (Pelargonic acid) |
| CAS | 112-05-0 |
| Purity Grade Notes | GC |
| Molecular Formula | C9 H18 O2 |
| Quantity | 25 g |
| Synonym | Nonanoic acid (7CI','8CI','9CI','ACI), 1-Octanecarboxylic acid, Emerion W 90PA, Emery 1202, Emery 1203, Grantrico, n-Nonanoic acid, n-Nonoic acid, n-Nonylic acid, n-Pelargonic acid, Nonoic acid, Nonylic acid, NSC 62787, NSC 65450, NSC 65455, Pelargonic acid, Prifrac 2914, Thinex |
| InChI Formula | InChI=1S/C9H18O2/c1-2-3-4-5-6-7-8-9(10)11/h2-8H2,1H3,(H,10,11) |
| SMILES | CCCCCCCCC(=O)O |
| IUPAC Name | nonanoic acid |
| Molecular Weight (g/mol) | 158.24 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?