missing translation for 'onlineSavingsMsg'
Learn More

n-Nonane, 99%, pure

Product Code. 10123610 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
2mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10123610 500 mL Glass bottle 500mL
10001420 2 mL Glass bottle 2mL
10153560 100 mL Glass bottle 100mL
10052633 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 10123610 Supplier Thermo Scientific Acros Supplier No. 129115000
  • ONLINE EXCLUSIVE PRICE
    £102.00
    £93.80 / 500mL
    Save £8.20
    8% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 111-84-2 | C9H20 | 128.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-84-2
MDL Number MFCD00009574
InChI Key BKIMMITUMNQMOS-UHFFFAOYSA-N
Synonym n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard
PubChem CID 8141
ChEBI CHEBI:32892
IUPAC Name nonane
SMILES CCCCCCCCC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -53.0°C
Color Colorless
Density 0.7180g/mL
Boiling Point 151.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H20
Linear Formula CH3(CH2)7CH3
Refractive Index 1.4040 to 1.4060
Quantity 500 mL
Beilstein 01, 165
Specific Gravity 0.718
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, slightly soluble in benzene, acetone, chloroform
Molecular Weight (g/mol) 128.26
Formula Weight 128.26
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material n-Nonane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Nonane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.