missing translation for 'onlineSavingsMsg'
Learn More

n-Nonadecane, 99%

Product Code. 11357555
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11357555

Brand: Thermo Scientific Alfa Aesar L05701.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 629-92-5 | C19H40 | 268.529 g/mol

n-Nonadecane is used as a fragrance agent in cosmetics and perfumes. It is used by egg parasitoid Trissolcus basalis to discriminate between female and male hosts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Nonadecane is used as a fragrance agent in cosmetics and perfumes. It is used by egg parasitoid Trissolcus basalis to discriminate between female and male hosts.

Solubility
Soluble in Chloroform, Hexane.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-92-5
Molecular Formula C19H40
Molecular Weight (g/mol) 268.529
MDL Number MFCD00009012
InChI Key LQERIDTXQFOHKA-UHFFFAOYSA-N
Synonym n-nonadecane, nonadekan, unii-nmy21d3y5t, nmy21d3y5t, unknown branched fragment of phospholipid, ch3-ch2 17-ch3, unknown phospholipid fragment, nonadecane, analytical standard, upl
PubChem CID 12401
ChEBI CHEBI:32927
IUPAC Name nonadecane
SMILES CCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 32°C to 34°C
Density 0.786
Boiling Point 330°C to 331°C
Flash Point 168°C (334°F)
Refractive Index 1.441
Quantity 100 g
Beilstein 1742892
Solubility Information Soluble in Chloroform,Hexane.
Formula Weight 268.53
Percent Purity 99%
Chemical Name or Material n-Nonadecane
Show More Show Less
compliance-icons
Product Identifier
  • Nonadecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : Nonadecane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.