missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosonebivolol, Mikromol

Product Code. 30460041
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30460041 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460041 Supplier Mikromol Supplier No. MM0947.040025
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Nitrosonebivolol, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitroso Nebivolol
CAS 1391051-68-5
Analyte or Component Names N-Nitroso Nebivolol
Shipping Condition Ice pack (-20°C)
Molecular Formula C22 H24 F2 N2 O5
Certifications/Compliance ISO 9001
InChI Formula InChI=1S/C22H24F2N2O5/c23-15-3-7-19-13(9-15)1-5-21(30-19)17(27)11-26(25-29)12-18(28)22-6-2-14-10-16(24)4-8-20(14)31-22/h3-4,7-10,17-18,21-22,27-28H,1-2,5-6,11-12H2
Solution Type Neat
SMILES OC(CN(CC(O)C1CCc2cc(F)ccc2O1)N=O)C3CCc4cc(F)ccc4O3
IUPAC Name N,N-bis[2-(6-fluorochroman-2-yl)-2-hydroxy-ethyl]nitrous amide
Molecular Weight (g/mol) 434.43
Quantity 25 mg
Formula Weight 434.1653 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.