missing translation for 'onlineSavingsMsg'
Learn More

N-(Nitrosomethyl)ethanolamine, TRC

Product Code. 30010000
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010000 50 mg 50mg
30004170 250 mg 250mg
18071530 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010000 Supplier Toronto Research Chemicals Supplier No. TRCN58905550MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26921-68-6
Molecular Formula C3H8N2O2
Molecular Weight (g/mol) 104.11
Synonym Ethanol, 2-(methylnitrosamino)- (6CI,7CI,8CI), 2-(Methylnitrosoamino)ethanol, Methyl(2-hydroxyethyl)nitrosamine, Methyl(2-hydroxyethyl)nitrosoamine, N-(Nitrosomethyl)ethanolamine, N-Methyl-N-nitrosoethanolamine, N-Nitrosomethyl(2-hydroxyethyl)amine
IUPAC Name N-(2-hydroxyethyl)-N-methylnitrous amide
SMILES CN(CCO)N=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C3H8N2O2/c1-5(4-7)2-3-6/h6H,2-3H2,1H3
Formula Weight 104.0586
Percent Purity >95%
Chemical Name or Material N-Nitroso-N-methylethanolamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.