missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosohexamethylenimine, TRC

Product Code. 30475929
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30475929 50 mg -
30468157 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30475929 Supplier Toronto Research Chemicals Supplier No. TRCN54658550MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 932-83-2
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.17
Synonym Hexahydro-1-nitroso-1H-azepine, N-Nitrosoazacycloheptane, N-Nitrosohexahydroazepine, Nitrosohexamethylenimine
IUPAC Name 1-nitrosoazepane
SMILES O=NN1CCCCCC1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H12N2O/c9-7-8-5-3-1-2-4-6-8/h1-6H2
Formula Weight 128.095
Chemical Name or Material N-Nitrosohexamethylenimine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.