missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodiisopropylamine (NdiPA), Mikromol™

Product Code. 18000920
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18000920 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18000920 Supplier Mikromol Supplier No. MM0877.790025
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Nitrosodiisopropylamine (NdiPA)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitrosodiisopropylamine
CAS 601-77-4
Analyte or Component Names N-Nitrosodiisopropylamine
Shipping Condition Room Temperature
Molecular Formula C6 H14 N2 O
Certifications/Compliance ISO 9001
Synonym 2-Propanamine, N-(1-methylethyl)-N-nitroso-, Diethylamine, 1,1'-dimethyl-N-nitroso- (6CI,7CI,8CI), N-(1-Methylethyl)-N-nitroso-2-propanamine, Diisopropyl-N-nitrosamine, Diisopropylnitrosamine, N-Nitrodiisopropylamine, N-Nitrosodiisopropylamine, NSC 336, Nitrosodiisopropylamine, NDIPA, DIPNA, NDPA
InChI Formula InChI=1S/C6H14N2O/c1-5(2)8(7-9)6(3)4/h5-6H,1-4H3
SMILES CC(C)N(N=O)C(C)C
IUPAC Name N,N-di(propan-2-yl)nitrous amide
Molecular Weight (g/mol) 130.188
Quantity 25 mg
Formula Weight 130.111 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.