missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodiisopropylamine (1mg/mL in Methanol), TRC

Product Code. 18070800
Change view
Click to view available options
Quantity:
1 x 1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18070800 1 x 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18070800 Supplier Toronto Research Chemicals Supplier No. TRCKIT89351X1ML
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 601-77-4
Molecular Formula C6 H14 N2 O
Quantity 1 x 1 mL
Synonym 2-Propanamine, N-(1-methylethyl)-N-nitroso-, Diethylamine, 1,1'-dimethyl-N-nitroso- (6CI,7CI,8CI), N-(1-Methylethyl)-N-nitroso-2-propanamine, Diisopropyl-N-nitrosamine, Diisopropylnitrosamine, N-Nitrodiisopropylamine, N-Nitrosodiisopropylamine, NSC 336, Nitrosodiisopropylamine, NDIPA, DIPNA, NDPA
InChI Formula InChI=1S/C6H14N2O/c1-5(2)8(7-9)6(3)4/h5-6H,1-4H3
SMILES CC(C)N(N=O)C(C)C
IUPAC Name N,N-di(propan-2-yl)nitrous amide
Molecular Weight (g/mol) 130.188
Formula Weight 130.111
Concentration or Composition (by Analyte or Components) N-Nitrosodiisopropylamine 1 mg/mL
Chemical Name or Material N-Nitrosodiisopropylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.