missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodiethylamine 0.1 mg/ml in Methanol, Mikromol™

Product Code. 18091000
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18091000 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18091000 Supplier Mikromol Supplier No. MM0487.10AME00.10
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Nitrosodiethylamine 0.1 mg/ml in Methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitroso-diethylamine
CAS 55-18-5
Analyte or Component Names N-Nitroso-diethylamine
Shipping Condition Ice pack (-20°C)
Molecular Formula C4 H10 N2 O
Certifications/Compliance ISO 9001
Synonym Ethanamine, N-ethyl-N-nitroso-, Diethylamine, N-nitroso- (6CI,8CI), N-Ethyl-N-nitrosoethanamine, DEN, DEN (mutagen), DENA, Diethylnitrosamide, Diethylnitrosamine, Diethylnitrosoamine, N,N-Diethylnitrosoamine, N-Nitroso-N,N-diethylamine, N-Nitrosodiethylamine, NDEA, NSC 132, Nitrosodiethylamine, N-Nitroso-diethylamine
InChI Formula InChI=1S/C4H10N2O/c1-3-6(4-2)5-7/h3-4H2,1-2H3
SMILES CCN(CC)N=O
IUPAC Name N,N-diethylnitrous amide
Solvent or Matrix Methanol
Molecular Weight (g/mol) 102.14
Quantity 1 mL
Formula Weight 102.0793 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.