missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodicyclohexylamine, TRC

Product Code. 17933295
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17933295 250 mg 250mg
17915095 50 mg 50mg
17913605 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17933295 Supplier Toronto Research Chemicals Supplier No. TRCN525550250MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947-92-2
Molecular Formula C12 H22 N2 O
Molecular Weight (g/mol) 210.32
Synonym N-Nitrosodicyclohexylamine, Dicyclohexylamine, N-nitroso- (6CI,7CI,8CI), N-Cyclohexyl-N-nitrosocyclohexanamine, Dicyclohexylnitrosamine, N,N-Dicyclohexylnitrosamine, NSC 337
IUPAC Name N,N-dicyclohexylnitrous amide
SMILES O=NN(C1CCCCC1)C2CCCCC2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H22N2O/c15-13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h11-12H,1-10H2
Formula Weight 210.1732
Percent Purity >95%
Chemical Name or Material N-Nitrosodicyclohexylamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.