missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodibenzylamine-d10 (1 mg/mL in Methanol), TRC

Product Code. 30390022
Change view
Click to view available options
Quantity:
1x1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390022 1x1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390022 Supplier Toronto Research Chemicals Supplier No. TRCKIT34911X1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitrosodibenzylamine D10 (Diphenyl D10)
CAS 1794791-25-5
Molecular Formula C14 D10 H4 N2 O
Quantity 1x1 mL
Synonym N-Nitrosodibenzylamine D10 (Diphenyl D10)
InChI Formula InChI=1S/C14H14N2O/c17-15-16(11-13-7-3-1-4-8-13)12-14-9-5-2-6-10-14/h1-10H,11-12H2/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D
SMILES [2H]c1c([2H])c([2H])c(CN(Cc2c([2H])c([2H])c([2H])c([2H])c2[2H])N=O)c([2H])c1[2H]
IUPAC Name N,N-bis[(2,3,4,5,6-pentadeuteriophenyl)methyl]nitrous amide
Molecular Weight (g/mol) 236.335
Formula Weight 236.173
Concentration or Composition (by Analyte or Components) 1 mg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.