missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodesipramine, TRC

Product Code. 30041351
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041351 50 mg 50mg
30040726 10 mg 10mg
30007382 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041351 Supplier Toronto Research Chemicals Supplier No. TRCN13739050MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57164-17-7
Molecular Formula C18H21N3O
Molecular Weight (g/mol) 295.38
Synonym 10,11-Dihydro-N-methyl-N-nitroso-5H-dibenz[b,f]azepine-5-propanamine
IUPAC Name N-[3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)propyl]-N-methylnitrous amide
SMILES CN(CCCN1c2ccccc2CCc3ccccc13)N=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H21N3O/c1-20(19-22)13-6-14-21-17-9-4-2-7-15(17)11-12-16-8-3-5-10-18(16)21/h2-5,7-10H,6,11-14H2,1H3
Formula Weight 295.1685
Chemical Name or Material N-Nitrosodesipramine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.