missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosobis(2-hydroxypropyl)amine, TRC

Product Code. 18061110
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18061110 100 mg 100mg
18003020 25 mg 25mg
18013420 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18061110 Supplier Toronto Research Chemicals Supplier No. TRCN525225100MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53609-64-6
Molecular Formula C6 H14 N2 O3
Molecular Weight (g/mol) 162.19
Synonym 1,1'-(Nitrosoimino)bis[2-propanol], 2,2'-Dihydroxydipropylnitrosamine, DHPN, Di(2-hydroxypropyl)nitrosamine, Diisopropanolnitrosamine, N-Bis(2-hydroxypropyl)nitrosamine, N-Nitrosobis(2-hydroxypropyl)amine, N-Nitrosodiisopropanolamine, Nitrosobis(2-hydroxypropyl)amine
IUPAC Name N,N-bis(2-hydroxypropyl)nitrous amide
SMILES CC(O)CN(CC(C)O)N=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H14N2O3/c1-5(9)3-8(7-11)4-6(2)10/h5-6,9-10H,3-4H2,1-2H3
Formula Weight 162.1004
Percent Purity >95%
Chemical Name or Material N-Nitrosobis(2-hydroxypropyl)amine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.