missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso Nebivolol (Mixture of Diastereomers), TRC

Product Code. 30133566
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133566 5 mg 5mg
17913865 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133566 Supplier Toronto Research Chemicals Supplier No. TRCN5319005MG
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1391051-68-5
Molecular Formula C22 H24 F2 N2 O5
Molecular Weight (g/mol) 434.43
Synonym N,N-Bis[2-(6-fluorochroman-2-yl)-2-hydroxyethyl]nitrous amide
IUPAC Name N,N-bis[2-(6-fluoro-3,4-dihydro-2H-chromen-2-yl)-2-hydroxyethyl]nitrous amide
SMILES OC(CN(CC(O)C1CCc2cc(F)ccc2O1)N=O)C3CCc4cc(F)ccc4O3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C22H24F2N2O5/c23-15-3-7-19-13(9-15)1-5-21(30-19)17(27)11-26(25-29)12-18(28)22-6-2-14-10-16(24)4-8-20(14)31-22/h3-4,7-10,17-18,21-22,27-28H,1-2,5-6,11-12H2
Formula Weight 434.1653
Percent Purity >95%
Chemical Name or Material N-Nitroso Nebivolol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.