missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso-N-methyl-3-aminopropionic Acid, TRC

Product Code. 30136310
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136310 25 mg 25mg
30135975 100 mg 100mg
30011538 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136310 Supplier Toronto Research Chemicals Supplier No. TRCN52920025MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10478-42-9
Molecular Formula C4 H8 N2 O3
Molecular Weight (g/mol) 132.12
Synonym Propanoic acid, 3-(methylnitrosoamino)- (ACI), 3-(Methylnitrosoamino)propanoic acid (ACI), β-Alanine, N-methyl-N-nitroso- (7CI, 8CI, 9CI), 3-(N-Nitroso-N-methylamino)propionic acid, NMPA, N-Nitroso-N-methyl-3-aminopropionic acid, N-Methyl-N-nitroso-β-alanine, 3-(N-Nitroso-N-methylamino)propanoic acid
IUPAC Name 3-[methyl(nitroso)amino]propanoic acid
SMILES CN(CCC(=O)O)N=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C4H8N2O3/c1-6(5-9)3-2-4(7)8/h2-3H2,1H3,(H,7,8)
Formula Weight 132.0535
Percent Purity >95%
Chemical Name or Material N-Nitroso-N-methyl-3-aminopropionic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.