missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso-N-ethylbutylamine, TRC

Product Code. 30132459
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132459 25 mg 25mg
30009693 10 mg 10mg
30010213 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132459 Supplier Toronto Research Chemicals Supplier No. TRCN52581025MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4549-44-4
Molecular Formula C6 H14 N2 O
Molecular Weight (g/mol) 130.19
Synonym N-Nitroso-N-ethylbutylamine, 1-Butanamine, N-ethyl-N-nitroso- (9CI, ACI), Butylamine, N-ethyl-N-nitroso- (7CI, 8CI), N-Ethyl-N-nitroso-1-butanamine (ACI), Butylethylnitrosamine, Ethyl-n-butylnitrosamine, Ethylbutylnitrosamine, N-Butyl-N-ethylnitrosamine, N-Ethyl-N-butylnitrosamine, N-Ethyl-N-nitrosobutylamine, N-Nitroso-N-butyl-N-ethylamine, N-Nitrosoethyl-n-butylamine, Nitrosobutylethylamine, NSC 135
IUPAC Name N-butyl-N-ethylnitrous amide
SMILES CCCCN(CC)N=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H14N2O/c1-3-5-6-8(4-2)7-9/h3-6H2,1-2H3
Formula Weight 130.1106
Chemical Name or Material N-Nitroso-N-ethylbutylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.