missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso-N-ethyl-N-isopropylamine 1.0 mg/ml in Methanol, Mikromol

Product Code. 30459299
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30459299 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459299 Supplier Mikromol Supplier No. MM0487.17L21000
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Nitroso-N-ethyl-N-isopropylamine 1.0 mg/ml in Methanol, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Ethyl-N-nitroso-2-propanamine
CAS 16339-04-1
Analyte or Component Names N-Ethyl-N-nitroso-2-propanamine
Shipping Condition Ice pack (-20°C)
Molecular Formula C5 H12 N2 O
Concentration 1 mg/mL
Certifications/Compliance ISO 9001
Synonym 2-Propanamine, N-ethyl-N-nitroso-, Diethylamine, 1-methyl-N-nitroso- (7CI,8CI), N-Ethyl-N-nitroso-2-propanamine, 1-Methyl-N-nitrosodiethylamine, Ethylisopropylnitrosamine
InChI Formula InChI=1S/C5H12N2O/c1-4-7(6-8)5(2)3/h5H,4H2,1-3H3
Solution Type Single Solution
SMILES CCN(N=O)C(C)C
IUPAC Name N-ethyl-N-propan-2-ylnitrous amide
Solvent or Matrix Methanol
Molecular Weight (g/mol) 116.1616
Quantity 1 mL
Formula Weight 116.095 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.