missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso Iminostilbene, TRC

Product Code. 30135217
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135217 25 mg 25mg
30133694 10 mg 10mg
30008904 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135217 Supplier Toronto Research Chemicals Supplier No. TRCN52810025MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38652-29-8
Molecular Formula C14H10N2O
Molecular Weight (g/mol) 222.24
Synonym 5-Nitroso-5H-dibenz[b,f]azepine;
IUPAC Name 11-nitrosobenzo[b][1]benzazepine
SMILES O=NN1c2ccccc2C=Cc3ccccc13

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H10N2O/c17-15-16-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)16/h1-10H
Formula Weight 222.0793
Chemical Name or Material N-Nitroso Iminostilbene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.