missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso Imidapril, TRC

Product Code. 30391371
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391371 5 mg 5mg
30402353 25 mg 25mg
30408380 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391371 Supplier Toronto Research Chemicals Supplier No. TRCI2750805MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitroso Imidapril
Analyte or Component Names N-Nitroso Imidapril
Shipping Condition Room Temperature
Molecular Formula C20 H26 N4 O7
Synonym (S)-3-(N-((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)-N-nitroso-L-alanyl)-1-methyl-2-oxoimidazolidine-4-carboxylic Acid
InChI Formula InChI=1S/C20H26N4O7/c1-4-31-19(28)15(11-10-14-8-6-5-7-9-14)24(21-30)13(2)17(25)23-16(18(26)27)12-22(3)20(23)29/h5-9,13,15-16H,4,10-12H2,1-3H3,(H,26,27)/t13-,15-,16-/m0/s1
Solution Type Neat
SMILES CCOC(=O)[C@H](CCc1ccccc1)N(N=O)[C@@H](C)C(=O)N2[C@@H](CN(C)C2=O)C(=O)O
IUPAC Name (4S)-3-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]-nitrosoamino]propanoyl]-1-methyl-2-oxoimidazolidine-4-carboxylic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 434.443
Quantity 5 mg
Formula Weight 434.18
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.