missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso-ethyl-isopropylamine D5 (ethyl D5), Dr. Ehrenstorfer

Product Code. 30315854
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315854 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30315854 missing translation for 'mfr' Dr. Ehrenstorfer missing translation for 'supplierNo' DRECA15605110
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitroso-ethyl-isopropylamine D5 (Ethyl D5)
CAS 2975202-10-7
Molecular Formula C5 2H5 H7 N2 O
Quantity 10 mg
Synonym N-Nitroso-ethyl-isopropylamine D5 (Ethyl D5), N-Ethyl-N-nitroso-2-propanamine-d5
InChI Formula InChI=1S/C5H12N2O/c1-4-7(6-8)5(2)3/h5H,4H2,1-3H3/i1D3,4D2
SMILES [2H]C([2H])([2H])C([2H])([2H])N(N=O)C(C)C
IUPAC Name N-(1,1,2,2,2-pentadeuterioethyl)-N-propan-2-ylnitrous amide
Molecular Weight (g/mol) 121.1924
Formula Weight 121.1263 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.