missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitroso Enalapril, TRC
Specifications
Specifications
| Chemical Name or Material | N-Nitrosoenalapril |
| Analyte or Component Names | N-Nitrosoenalapril |
| Purity Grade Notes | HPLC |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H27 N3 O6 |
| Synonym | N-Nitroso Enalapril, (2S)-1-[(2S)-2-[[(1S)-1-Ethoxycarbonyl-3-phenyl-propyl]-nitroso-amino]propanoyl]pyrrolidine-2-carboxylic acid, N-Nitrosoenalapril |
| InChI Formula | InChI=1S/C20H27N3O6/c1-3-29-20(27)17(12-11-15-8-5-4-6-9-15)23(21-28)14(2)18(24)22-13-7-10-16(22)19(25)26/h4-6,8-9,14,16-17H,3,7,10-13H2,1-2H3,(H,25,26)/t14-,16-,17-/m0/s1 |
| Solution Type | Neat |
| SMILES | CCOC(=O)[C@H](CCc1ccccc1)N(N=O)[C@@H](C)C(=O)N2CCC[C@H]2C(=O)O |
| IUPAC Name | (2S)-1-[(2S)-2-[[(1S)-1-ethoxycarbonyl-3-phenyl-propyl]-nitroso-amino]propanoyl]pyrrolidine-2-carboxylic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?