missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso-di-n-butylamine, TRC

Product Code. 30136340
Change view
Click to view available options
Quantity:
10 mg
100 mg
5 g
Unit Size:
100mg
10mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136340 5 g 5g
18053920 100 mg 100mg
17979619 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136340 Supplier Toronto Research Chemicals Supplier No. TRCN5255355G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 924-16-3
Molecular Formula C8 H18 N2 O
Molecular Weight (g/mol) 158.24
Synonym 1-Butanamine, N-butyl-N-nitroso-, Dibutylamine, N-nitroso- (6CI,7CI,8CI), N-Butyl-N-nitroso-1-butanamine, Di-n-butylnitrosamine, Dibutylnitrosamine, N,N-Di-n-butylnitrosamine, N,N-Dibutylnitrosamine, N-Nitroso-N-di-n-butylamine, N-Nitrosodi-n-butylamine, N-Nitrosodibutylamine, NDBA, NSC 6830, Nitrosodibutylamine, N-Nitroso-di-n-butylamine
IUPAC Name N,N-dibutylnitrous amide
SMILES CCCCN(CCCC)N=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H18N2O/c1-3-5-7-10(9-11)8-6-4-2/h3-8H2,1-2H3
Formula Weight 158.1419
Percent Purity >95%
Chemical Name or Material N-Nitroso-di-n-butylamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.