missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso Desloratadine, TRC

Product Code. 30483359
Change view
Click to view available options
Quantity:
5 mg
50 mg
Product Code. Quantity
30483359 50 mg
30468466 5 mg
2 options
This item is not returnable. View return policy

Product Code. 30483359

Brand: Toronto Research Chemicals TRCN52537050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246819-22-6
Formula Weight 339.1138
InChI Formula InChI=1S/C19H18ClN3O/c20-16-5-6-17-15(12-16)4-3-14-2-1-9-21-19(14)18(17)13-7-10-23(22-24)11-8-13/h1-2,5-6,9,12H,3-4,7-8,10-11H2
IUPAC Name 8-chloro-11-(1-nitrosopiperidin-4-ylidene)-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridine
Molecular Formula C19 H18 Cl N3 O
Molecular Weight (g/mol) 339.8187
Percent Purity >95%
SMILES Clc1ccc2C(=C3CCN(CC3)N=O)c4ncccc4CCc2c1
Synonym 5H-Benzo[5,6]cyclohepta[1,2-b]pyridine, 8-chloro-6,11-dihydro-11-(1-nitroso-4-piperidinylidene)-, 8-Chloro-6,11-dihydro-11-(1-nitroso-4-piperidinylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine, N-Nitroso desloratadine, N-Nitrosodesloratadine
Chemical Name or Material 8-Chloro-6,11-dihydro-11-(1-nitroso-4-piperidinylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
Grade HPLC
Show More Show Less

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.