missing translation for 'onlineSavingsMsg'
Learn More

N-n-Tetradecylmethylamine, TRC

Product Code. 30420586
Change view
Click to view available options
Quantity:
25 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30420586 25 mg
30423508 250 mg
30414702 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30420586 Supplier Toronto Research Chemicals Supplier No. TRCT29263025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29369-63-9
Molecular Formula C15H33N
Molecular Weight (g/mol) 227.43
Synonym n-Tetradecylmethylamine, N-Methyltetradecanamine, N-Methyltetradecylamine, N-Methyltetradecylamine, N-Methyl-1-tetradecanamine
IUPAC Name N-methyltetradecan-1-amine
SMILES CCCCCCCCCCCCCCNC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H33N/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-2/h16H,3-15H2,1-2H3
Formula Weight 227.2613
Chemical Name or Material N-n-Tetradecylmethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.