missing translation for 'onlineSavingsMsg'
Learn More

N-(n-Propyl)thiourea, 98%

Product Code. 11398255
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11398255

Brand: Thermo Scientific Alfa Aesar L10149.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 927-67-3 | C4H10N2S | 118.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 927-67-3
Molecular Formula C4H10N2S
Molecular Weight (g/mol) 118.20
MDL Number MFCD00041195
InChI Key UHGKYJXJYJWDAM-UHFFFAOYSA-N
Synonym n-propylthiourea, 1-propyl-2-thiourea, 1-propylthiourea, propyl-2-thiourea, thiourea, propyl, thiourea, n-propyl, propyl isothiourea, thiourea,n-propyl, n-n-propyl thiourea, n-prop-2-en-1-ylthioformamide
PubChem CID 1615208
IUPAC Name propylthiourea
SMILES CCCNC(N)=S

Specifications

Melting Point 109°C to 112°C
Odor Odorless
Quantity 5 g
UN Number UN2811
Beilstein 1743064
Formula Weight 118.2
Percent Purity 98%
Chemical Name or Material N-(n-Propyl)thiourea

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.